Learn More
Saint-Gobain AstiPure™ RF1 Series PFA Male Tube Connector
Use these fittings to deliver corrosive fluids. Saint Gobain Performance Plastics™ AstiPure™ RF1 Series PFA Male Tube Connector are leak proof and reliable under a wide range of temperatures and pressures.
Brand: Saint-Gobain RF10413
Additional Details : Weight : 0.01800kg
Description
- Sturdy rubber Silicone piston rings for flasks
- For general lab applications
Specifications
Male Tube Connector | |
40 mm | |
1 Each | |
1/4 in. NPT |
PFA | |
For 4 x 6mm dia. tubing | |
13mm |