Learn More
Saint-Gobain AstiPure™ RVT1 Series PFA Flare Adapter
Brand: Saint-Gobain RVT10408
Additional Details : Weight : 0.80000kg
Specifications
Flare Adapter | |
PFA | |
1 Each |
8 mm | |
For 4 x 6mm tubing O.D. |