Läs mer
Astragaloside I, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Astragaloside I |
| CAS | 84680-75-1 |
| Molekylformel | C45 H72 O16 |
| Kvantitet | 5 mg |
| Synonym | β-D-Glucopyranoside, (3β,6α,16β,20R,24S)-3-[(2,3-di-O-acetyl-β-D-xylopyranosyl)oxy]-20,24-epoxy-16,25-dihydroxy-9,19-cyclolanostan-6-yl, 1H,19H-Cyclopropa[9,10]cyclopenta[a]phenanthrene, β-D-glucopyranoside deriv., 9,19-Cyclolanostane, β-D-glucopyranoside deriv., (3β,6α,16β,20R,24S)-3-[(2,3-Di-O-acetyl-β-D-xylopyranosyl)oxy]-20,24-epoxy-16,25-dihydroxy-9,19-cyclolanostan-6-yl β-D-glucopyranoside, Astragaloside I, Astrasieversianin IV, Cyclosieversioside B |
| InChI formel | InChI=1S/C45H72O16/c1-21(47)56-33-24(50)19-55-38(34(33)57-22(2)48)60-28-11-13-45-20-44(45)15-14-41(7)35(43(9)12-10-29(61-43)40(5,6)54)23(49)17-42(41,8)27(44)16-25(36(45)39(28,3)4)58-37-32(53)31(52)30(51)26(18-46)59-37/h23-38,46,49-54H,10-20H2,1-9H3/t23-,24+,25-,26+,27-,28-,29-,30+,31-,32+,33-,34+,35-,36-,37+,38-,41+,42-,43+,44-,45+/m0/s1 |
| LEDER | CC(=O)O[C@H]1[C@H](O)CO[C@@H](O[C@H]2CC[C@]34C[C@]35CC[C@]6(C)[C@H]([C@@H](O)C[C@@]6(C)[C@@H]5C[C@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H]4C2(C)C)[C@@]8(C)CC[C@H](O8)C(C)(C)O)[C@@H]1OC(=O)C |
| Molekylvikt (g/mol) | 869.04 |
| Formel vikt | 868.482 |