Läs mer
Astragaloside IV, TRC
Produktkod. 30134947
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134947 10 mg
30006201 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134947 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79013010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84687-43-4
Molekylformel C41 H68 O14
Molekylvikt (g/mol) 784.97
Synonym (3β,6α,16β,20R,24S)-20,24-Epoxy-16,25-dihydroxy-3-(β-D-xylopyranosyloxy)-9,19-cyclolanostan-6-yl β-D-glucopyranoside, 1H,19H-Cyclopropa[9,10]cyclopenta[a]phenanthrene, β-D-glucopyranoside deriv., 9,19-Cyclolanostane, β-D-glucopyranoside deriv., Astragaloside IV, Astrasieversianin XIV, Astraversianin XIV, Cyclosieversioside F, Cyclosiversioside F
LEDER CC(C)(O)[C@@H]1CC[C@@](C)(O1)[C@H]2[C@@H](O)C[C@@]3(C)[C@@H]4C[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)[C@H]6C(C)(C)[C@H](CC[C@@]67C[C@@]47CC[C@]23C)O[C@@H]8OC[C@@H](O)[C@H](O)[C@H]8O
Specifikationer
Kemiskt namn eller material Astragaloside IV
Analyt- eller komponentnamn Astragaloside IV
Frakt skick Room Temperature
InChI formel InChI=1S/C41H68O14/c1-35(2)24(54-33-29(48)26(45)20(44)17-51-33)9-11-41-18-40(41)13-12-37(5)31(39(7)10-8-25(55-39)36(3,4)50)19(43)15-38(37,6)23(40)14-21(32(35)41)52-34-30(49)28(47)27(46)22(16-42)53-34/h19-34,42-50H,8-18H2,1-7H3/t19-,20+,21-,22+,23-,24-,25-,26-,27+,28-,29+,30+,31-,32-,33-,34+,37+,38-,39+,40-,41+/m0/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 10 mg
Formel vikt 784.4609