Läs mer
Asulam, TRC
Produktkod. 30347189
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30347189 500 mg
1 options
Produktkod. 30347189 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA788750500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Asulam
CAS 3337-71-1
Molekylformel C8 H10 N2 O4 S
Kvantitet 500 mg
Synonym Carbamic acid, [(4-aminophenyl)sulfonyl]-, methyl ester (9CI), Carbamic acid, sulfanilyl-, methyl ester (7CI,8CI), Asilan, Asulam, Asulox F, M and B 9057, MB 9057, Methyl (4-aminophenylsulfonyl)carbamate, Methyl 4-aminobenzenesulfonylcarbamate, Methyl p-aminobenzenesulfonylcarbamate, Methyl sulfanilylcarbamate, N1-Methoxycarbonylsulfanilamide, Plakin, Plakin (herbicide), Rattler, Rattler (herbicide)
InChI formel InChI=1S/C8H10N2O4S/c1-14-8(11)10-15(12,13)7-4-2-6(9)3-5-7/h2-5H,9H2,1H3,(H,10,11)
LEDER COC(=O)NS(=O)(=O)c1ccc(N)cc1
IUPAC-namn methyl N-(4-aminophenyl)sulfonylcarbamate
Molekylvikt (g/mol) 230.24
Formel vikt 230.0361