missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ASXL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578646
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Positive Control: human ASXL1-transfected 293T cells Predicted Reactivity: Mouse (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probable Polycomb group (PcG) protein involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as retinoic acid receptors (RARs) and peroxisome proliferator-activated receptor gamma (PPARG). Acts as coactivator of RARA and RXRA through association with NCOA1. Acts as corepressor through recruitment of KDM1A and CBX5 to target genes in a cell-type specific manner; the function seems to involve differential recruitment of methylated histone H3 to respective promoters. Acts as corepressor for PPARG and suppresses its adipocyte differentiation-inducing activity. Non-catalytic component of the PR-DUB complex, a complex that specifically mediates deubiquitination of histone H2A monoubiquitinated at 'Lys-119' (H2AK119ub1).
Specifikationer
| ASXL1 | |
| Polyclonal | |
| Unconjugated | |
| Asxl1 | |
| additional sex combs like 1; additional sex combs like 1, transcriptional regulator; additional sex combs like transcriptional regulator 1; additional sex combs-like protein 1; Asxl1; BOPS; KIAA0978; MDS; MGC117280; MGC71111; mKIAA0978; Polycomb group protein ASXL1; putative Polycomb group protein ASXL1; RGD1561878 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171023, 228790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P59598, Q8IXJ9 | |
| Asxl1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human ASXL1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering