Läs mer
AT7867 dihydrochloride, MedChemExpress
Produktkod. 16481207
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16481207 50 mg -
16477697 5 mg 5mg
16465736 100 mg 100mg
16404968 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16481207 Leverantör Medchem Express Leverantörsnummer HY12059A
MedChemExpress AT7867 dihydrochloride is a potent ATP-competitive inhibitor of Akt1/Akt2/Akt3 and p70S6K/PKA with IC50s of 32 nM/17 nM/47 nM and 85 nM/20 nM, respectively.
AT7867 dihydrochloride is a potent ATP-competitive inhibitor of Akt1/Akt2/Akt3 and p70S6K/PKA with IC50s of 32 nM/17 nM/47 nM and 85 nM/20 nM, respectively.
Specifikationer
Kemiskt namn eller material AT7867 dihydrochloride
CAS 1431697-86-7
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C20H22Cl3N3
Kvantitet 50 mg
Löslighetsinformation DMSO : 50 mg/mL (121.72 mM; Need ultrasonic) ∣H2O : 2.27 mg/mL (5.53 mM; ultrasonic and warming and heat to 60°C)
LEDER ClC1=CC=C(C=C1)C2(CCNCC2)C3=CC=C(C=C3)C4=CNN=C4.[H]Cl.[H]Cl
Molekylvikt (g/mol) 410.77
Formel vikt 410.77
Procent renhet 99.17%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease