Description
- Automatic Temperature Compensation function
- Brix
- Inverted sugar concentration
- HFCS-55 ( High Fructose Corn Syrup )
- HFCS-42 ( High Fructose Corn Syrup )
Services
Two year limited warranty against manufacturer defects from the date of the original purchase. Warranty does not cover or apply to the prism. Period can be extended to three years if the product is registered with ATAGO.
Specifications
Refractometer | |
1.32700 to 1.58000 | |
Refractive Index: ±0.00004; Brix: ±0.03% | |
Brix, Refractive Index |
0.00 to 100.00 Brix | |
SMART-1 | |
Min. Indication: 0.00001%; Brix: 0.01% | |
5°C to 60°C |