Learn More
ATAGO™ Digital EC Meter, Model DEC-2
Ideal for soil and hydroponics, industrial washing liquids, checking wastewater and other soluble solutions. The Atago™ DEC-2 Digital EC Meter features Automatic Temperature Compensation from 0° to 50°C for measuring conductivity.
Brand: ATAGO™ 4340
Additional Details : Weight : 0.01000kg
Description
- Compact and easy to carry
- Calibration with exclusive standard liquid
- One-handed operation
- Waterproof design
- Battery operated
- Digital display
Specifications
DEC-2 | |
0.10 ms/cm | |
Digital EC Meter |
Watch battery (LR44) 1.5V – 4 | |
0.0°C to 50.0°C | |
0.00 to 19.90 ms/cm |