Learn More
ATAGO™ Digital Urine Specific Gravity Refractometer, UG-α
The ATAGO™ Digital Urine Specific Gravity Refractometer UG-α is an upgraded version of previous model UG-1. The minimum scale indication has been improved to display up to the fourth decimal.
Brand: ATAGO™ 3464
Additional Details : Weight : 0.27000kg
Description
- Using the principles of light refraction, ATAGO Clinical Refractometers use special scales for clinical purposes such as specific gravity of urine
- After pressing the START key, with the sample placed on the prism, the measurement values will be displayed on the LCD streaming as an electric message board
Specifications
Refractometer | |
UG-α | |
Min. Indication: 0.0001 | |
10°C to 35°C |
1.0000 to 1.0600 | |
± 0.0010 | |
Urine Specific Gravity |