Learn More
Abnova™ ATF1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FS0048.100uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16023485
  • 15410.00 SEK / 100µL
Estimated Shipment: 08-11-2023
to see stock.
Specifications
466
EWS-ATF1,FUS/ATF-1,TREB36
Fluorescent In Situ Hybridization (Cell)
100 μL
ATF1
Store at 4°C in the dark
activating transcription factor 1
DNA/RNA