missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | ATG16L1 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879294
|
Proteintech
19812-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16869294
|
Proteintech
19812-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Autophagy related protein 16 L is encoded by the ATG16L gene and is located on chromosome 2 in humans. It is also known as WD30 or IBD10 (Inflammatory Bowel Disease 10). ATG 12 and ATG5 conjugate with each other and undergoes homo oligomerisation with ATG16L to form an 800 kDa complex called the Atg16L complex. This complex facilitates coupling or lipidation of LC3 to phosphatidylethanolamine (PE) and this ubiquitin-like conjugation system is a widely used marker for autophagy. However, the ATG 16L complex along with other ATGs like ATG 18 and ATG 21 prevent the premature cleavage of ATG8-PE by the cysteine protease, Atg4 until the autophagosome is deemed complete.Specifications
ATG16L1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q676U5, Q8C0J2 | |
55054, 77040 | |
ATG16L1 Fusion Protein Ag13844 | |
Primary | |
-20°C | |
ATG16L1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
APG16 like 1, APG16L, ATG16L, ATG16L1, Autophagy related protein 16 1, IBD10, WDR30 | |
ATG16L1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |