Läs mer
Athidathion, TRC
Produktkod. 30327136
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30327136 100 mg
1 options
Produktkod. 30327136 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA291180100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Athidathion
CAS 19691-80-6
Molekylformel C8 H15 N2 O4 P S3
Kvantitet 100 mg
Synonym Phosphorodithioic acid, O,O-diethyl ester, S-ester with 4-(mercaptomethyl)-2-methoxy-δ2-1,3,4-thiadiazolin-5-one (7CI,8CI), δ2-1,3,4-Thiadiazolin-5-one, 4-(mercaptomethyl)-2-methoxy-, S-ester with O,O-diethyl phosphorodithioate (8CI), Athidathion, GS 13006
InChI formel InChI=1S/C8H15N2O4PS3/c1-4-13-15(16,14-5-2)17-6-10-8(11)18-7(9-10)12-3/h4-6H2,1-3H3
LEDER CCOP(=S)(OCC)SCN1N=C(OC)SC1=O
IUPAC-namn 3-(diethoxyphosphinothioylsulfanylmethyl)-5-methoxy-1,3,4-thiadiazol-2-one
Molekylvikt (g/mol) 330.38
Formel vikt 329.9932