Läs mer
Tocris Bioscience™ ATI 2341
CXCR4 allosteric agonist
Brand: Tocris Bioscience™ 5795/1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ATI 2341 is a CXCR4 allosteric agonist (EC50 = 194 nM). Induces chemotaxis in CXCR4-expressing cells in vitro. Mobilizes polymorphonuclear neutrophils (PMNs) and hematopoietic stem and progenitor cells (HSPCs) from bone marrow niche in vivo. Exhibits biased signaling towards Gi proteins over G13 and β-arrestin.Specifikationer
| 1337878-62-2 | |
| Chemokine Receptor Agonists | |
| 1 mg |
| Store at -20°C | |
| Observed M.W.: 2256.82 | |
| C104H178N26O25S2 |