Läs mer
Atomoxetine Hydrochloride, TRC
Produktkod. 30134292
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134292 5 mg
30137422 100 mg
18000030 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134292 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7914005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82248-59-7
Molekylformel C17 H21 N O . Cl H
Molekylvikt (g/mol) 291.82
Synonym Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride (1:1), (γR)-, Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride, (R)-, Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride, (γR)- (9CI), (R)-(-)-N-Methyl-3-[(2-methylphenyl)oxy]-3-phenyl-1-aminopropane hydrochloride, (R)-(-)-Tomoxetine hydrochloride, (R)-N-Methyl-3-(2-methylphenoxy)-3-phenylpropylamine hydrochloride, (R)-Tomoxetine hydrochloride, Atomoxetine hydrochloride, LY 139603, Strattera, Tomoxetine hydrochloride
IUPAC-namn (3R)-N-methyl-3-(2-methylphenoxy)-3-phenylpropan-1-amine;hydrochloride
LEDER Cl.CNCC[C@@H](Oc1ccccc1C)c2ccccc2
Specifikationer
Kemiskt namn eller material Atomoxetine hydrochloride
Analyt- eller komponentnamn Atomoxetine hydrochloride
Frakt skick Room Temperature
InChI formel InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m1./s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 5 mg
Formel vikt 291.139