Läs mer
Atorvastatin-Amlodipine Adduct, TRC
Produktkod. 30341427
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30341427 25 mg
1 options
Produktkod. 30341427 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79169525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Atorvastatin-Amlodipine Adduct
CAS 425408-16-8
Molekylformel C53H58ClFN4O9
Kvantitet 25 mg
Synonym 3-Ethyl 5-Methyl 4-(2-Chlorophenyl)-2-[[2-[[(3R,5R)-7-[2-(4-fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]-3,5-dihydroxy-1-oxoheptyl]amino]ethoxy]methyl]-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylate
LEDER CCOC(=O)C1=C(COCCNC(=O)C[C@H](O)C[C@H](O)CCn2c(C(C)C)c(C(=O)Nc3ccccc3)c(c4ccccc4)c2c5ccc(F)cc5)NC(=C(C1c6ccccc6Cl)C(=O)OC)C
Molekylvikt (g/mol) 949.5
Formel vikt 948.3876