Läs mer
Atorvastatin Di-acetonide tert-Butyl Ester, TRC
Produktkod. 30363077
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363077 100 mg
30362268 50 mg
30361709 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363077 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD303481100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1116118-82-1
Molekylformel C50H64FN3O8
Molekylvikt (g/mol) 854.06
Synonym 1,1-Dimethylethyl (4R,6R)-6-[2-[[2-[(4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetyl]amino]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetate
LEDER CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H]5C[C@H](CC(=O)NCC[C@@H]6C[C@H](CC(=O)OC(C)(C)C)OC(C)(C)O6)OC(C)(C)O5
Specifikationer
Kemiskt namn eller material Atorvastatin Di-acetonide tert-Butyl Ester
Kvantitet 100 mg
InChI formel InChI=1S/C50H64FN3O8/c1-32(2)45-44(47(57)53-36-18-14-11-15-19-36)43(33-16-12-10-13-17-33)46(34-20-22-35(51)23-21-34)54(45)27-25-38-29-39(60-50(8,9)59-38)30-41(55)52-26-24-37-28-40(61-49(6,7)58-37)31-42(56)62-48(3,4)5/h10-23,32,37-40H,24-31H2,1-9H3,(H,52,55)(H,53,57)/t37-,38-,39-,40-/m1/s1
Formel vikt 853.4677