Läs mer
Atorvastatin Epoxydione Impurity, TRC
Produktkod. 17924775
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17924775 25 mg
17963115 10 mg
17923325 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17924775 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79185525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 148146-51-4
Molekylformel C26 H22 F N O4
Molekylvikt (g/mol) 431.46
Synonym 3-[(4-Fluorophenyl)carbonyl]-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide, Atorvastatin Calcium Trihydrate Imp. D (EP)
IUPAC-namn 3-(4-fluorobenzoyl)-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide
LEDER CC(C)C(=O)C1(OC1(C(=O)c2ccc(F)cc2)c3ccccc3)C(=O)Nc4ccccc4
Specifikationer
Kemiskt namn eller material 3-[(4-fluorophenyl)carbonyl]-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide
Analyt- eller komponentnamn 3-[(4-fluorophenyl)carbonyl]-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C26H22FNO4/c1-17(2)22(29)26(24(31)28-21-11-7-4-8-12-21)25(32-26,19-9-5-3-6-10-19)23(30)18-13-15-20(27)16-14-18/h3-17H,1-2H3,(H,28,31)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 25 mg
Formel vikt 431.1533
Procent renhet >95%