missing translation for 'onlineSavingsMsg'
Läs mer

Atorvastatin Lactam Phenanthrene Calcium Salt Impurity (mixture of diastereomers), TRC

Produktkod. 30333304
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30333304 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30333304 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7918155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (3R,5R)-7-[9-fluoro-2-oxo-3-(phenylcarbamoyl)-3-propan-2-ylphenanthro[9,10-b]pyrrol-1-yl]-3,5-dihydroxyheptanoate calcium salt
CAS 148127-12-2
Molekylformel 2 C33 H32 F N2 O6 . Ca
Kvantitet 5 mg
Synonym [3R-[1(βR*,δR*),3R*]]-9-Fluoro-2,3-dihydro-β,δ-dihydroxy-3-(1-methylethyl)-
InChI formel InChI=1S/2C33H33FN2O6.Ca/c2*1-19(2)33(31(41)35-21-8-4-3-5-9-21)29-25-11-7-6-10-24(25)27-16-20(34)12-13-26(27)30(29)36(32(33)42)15-14-22(37)17-23(38)18-28(39)40;/h2*3-13,16,19,22-23,37-38H,14-15,17-18H2,1-2H3,(H,35,41)(H,39,40);/q;;+2/p-2/t2*22-,23-,33?;/m11./s1
LEDER [Ca+2].CC(C)C1(C(=O)Nc2ccccc2)C(=O)N(CC[C@@H](O)C[C@@H](O)CC(=O)[O-])c3c1c4ccccc4c5cc(F)ccc35.CC(C)C6(C(=O)Nc7ccccc7)C(=O)N(CC[C@@H](O)C[C@@H](O)CC(=O)[O-])c8c6c9ccccc9c%10cc(F)ccc8%10
IUPAC-namn calcium;(3R,5R)-7-[9-fluoro-2-oxo-3-(phenylcarbamoyl)-3-propan-2-ylphenanthro[9,10-b]pyrrol-1-yl]-3,5-dihydroxyheptanoate
Molekylvikt (g/mol) 1183.31
Formel vikt 1182.4115
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.