Läs mer
Atorvastatin Lactam Phenanthrene Methyl Ester(Mixture of Diastereomers), TRC
Produktkod. 30343976
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30343976 10 mg
1 options
Produktkod. 30343976 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79188010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Atorvastatin Lactam Phenanthrene Methyl Ester(Mixture of Diastereomers)
CAS 906552-20-3
Molekylformel C34H35FN2O6
Kvantitet 10 mg
Synonym (βR,δR)-9-Fluoro-2,3-dihydro-β,δ-dihydroxy-3-(1-methylethyl)-2-oxo-3-[(phenylamino)carbonyl]-1H-dibenz[e,g]indole-1-heptanoic Acid Methyl Ester
InChI formel InChI=1S/C34H35FN2O6/c1-20(2)34(32(41)36-22-9-5-4-6-10-22)30-26-12-8-7-11-25(26)28-17-21(35)13-14-27(28)31(30)37(33(34)42)16-15-23(38)18-24(39)19-29(40)43-3/h4-14,17,20,23-24,38-39H,15-16,18-19H2,1-3H3,(H,36,41)/t23-,24-,34?/m1/s1
LEDER O[C@@H](C[C@@H](O)CC(OC)=O)CCN1C2=C(C(C=CC=C3)=C3C4=C2C=CC(F)=C4)C(C(NC5=CC=CC=C5)=O)(C(C)C)C1=O
IUPAC-namn methyl (3R,5R)-7-(9-fluoro-3-isopropyl-2-oxo-3-(phenylcarbamoyl)-2,3-dihydro-1H-dibenzo[e,g]indol-1-yl)-3,5-dihydroxyheptanoate
Molekylvikt (g/mol) 586.65
Formel vikt 586.25