Läs mer
Atosiban Acetate, TRC
Produktkod. 30135988
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135988 250 mg
17992625 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135988 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA791890250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 914453-95-5
Molekylformel C43 H67 N11 O12 S2 . C2 H4 O2
Molekylvikt (g/mol) 1054.24
Synonym Glycinamide, O-ethyl-N-(3-mercapto-1-oxopropyl)-D-tyrosyl-L-isoleucyl-L-threonyl-L-asparaginyl-L-cysteinyl-L-prolyl-L-ornithyl-, cyclic (1→5)-disulfide, acetate (1:1), Glycinamide, O-ethyl-N-(3-mercapto-1-oxopropyl)-D-tyrosyl-L-isoleucyl-L-threonyl-L-asparaginyl-L-cysteinyl-L-prolyl-L-ornithyl-, cyclic (1→5)-disulfide, monoacetate (salt) (9CI), Atosiban acetate
IUPAC-namn acetic acid;(2S)-N-[(2S)-5-amino-1-[(2-amino-2-oxoethyl)amino]-1-oxopentan-2-yl]-1-[(4R,7S,10S,13S,16R)-7-(2-amino-2-oxoethyl)-13-[(2S)-butan-2-yl]-16-[(4-ethoxyphenyl)methyl]-10-[(1R)-1-hydroxyethyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl]pyrrolidine-2-carboxamide
LEDER CCOc1ccc(C[C@H]2NC(=O)CCSSC[C@H](NC(=O)[C@H](CC(=O)N)NC(=O)[C@@H](NC(=O)[C@@H](NC2=O)[C@@H](C)CC)[C@@H](C)O)C(=O)N3CCC[C@H]3C(=O)N[C@@H](CCCN)C(=O)NCC(=O)N)cc1.CC(=O)O
Specifikationer
Kemiskt namn eller material Atosiban Acetate
Analyt- eller komponentnamn Atosiban Acetate
Frakt skick Room Temperature
InChI formel InChI=1S/C43H67N11O12S2.C2H4O2/c1-5-23(3)35-41(63)53-36(24(4)55)42(64)50-29(20-32(45)56)38(60)51-30(43(65)54-17-8-10-31(54)40(62)49-27(9-7-16-44)37(59)47-21-33(46)57)22-68-67-18-15-34(58)48-28(39(61)52-35)19-25-11-13-26(14-12-25)66-6-2;1-2(3)4/h11-14,23-24,27-31,35-36,55H,5-10,15-22,44H2,1-4H3,(H2,45,56)(H2,46,57)(H,47,59)(H,48,58)(H,49,62)(H,50,64)(H,51,60)(H,52,61)(H,53,63);1H3,(H,3,4)/t23-,24+,27-,28+,29-,30-,31-,35-,36-;/m0./s1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 250 mg
Formel vikt 1053.4623