missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MedChemExpress ATP
Specifikationer
Specifikationer
| Molekylvikt (g/mol) | 507.18 g/mol |
| Produkttyp | Natural Products |
| Innehåll och lagring | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Frakt skick | Room temperature |
| Renhet | 98.0% |
| Kvantitet | 500 mg |
| Löslighetsinformation | H2O : ≥ 100 mg/mL (197.17 mM) |
| För användning med (applikation) | COVID-19-immunoregulation |
| CAS | 56-65-5 |
| Synonym | Adenosine 5'-triphosphate |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?