Läs mer
ATP disodium salt, MedChemExpress
Produktkod. 16402149
Encompass_Preferred
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16402149 1 g
16450888 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16402149 Leverantör Medchem Express Leverantörsnummer HYB0345A
MedChemExpress ATP disodium salt (Adenosine 5'-triphosphate disodium salt) is a central component of energy storage and metabolism in vivo, provides the metabolic energy to drive metabolic pumps and serves as a coenzyme in cells. ATP disodium salt is an important endogenous signaling molecule in immunity and inflammation.
ATP disodium salt (Adenosine 5'-triphosphate disodium salt) is a central component of energy storage and metabolism in vivo, provides the metabolic energy to drive metabolic pumps and serves as a coenzyme in cells. ATP disodium salt is an important endogenous signaling molecule in immunity and inflammation.
Specifikationer
Kemiskt namn eller material ATP disodium salt
CAS 987-65-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C10H14N5Na2O13P3
Kvantitet 1 g
Synonym Adenosine 5'-triphosphate disodium salt Disodium adenosine triphosphate
Löslighetsinformation H2O : 100 mg/mL (181.44 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.81 mM; ultrasonic and warming and heat to 80°C)
LEDER O[C@@H]([C@H]([C@H](N1C=NC2=C1N=CN=C2N)O3)O)[C@H]3COP(O)(OP(OP([O-])(O)=O)([O-])=O)=O.[Na+].[Na+]
Molekylvikt (g/mol) 551.14
Formel vikt 551.14
Procent renhet 98.0%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Metabolism-protein/nucleotide metabolism