missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5F1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15999-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the b subunit of the proton channel.Specifications
ATP5F1 | |
Polyclonal | |
Unconjugated | |
ATP5F1 | |
ATP5F1, ATPase subunit b, PIG47 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11950, 171375, 515 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P19511, P24539, Q9CQQ7 | |
Atp5f1 | |
ATP5F1 Fusion Protein Ag8571 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |