Learn More
ATP5F1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15999-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16835434
  • 1656.73 SEK / 20µL
Estimated Shipment: 10-06-2024
to see stock.
Description
Description
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the b subunit of the proton channel.
Specifications
Specifications
ATP5F1
Polyclonal
Unconjugated
ATP5F1
ATP5F1, ATPase subunit b, PIG47
Rabbit
Antigen Affinity Chromatography
RUO
11950, 171375, 515
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P19511, P24539, Q9CQQ7
Atp5f1
ATP5F1 Fusion Protein Ag8571
20 μL
Primary
Human, Mouse, Rat