missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ ATP5G1/ATP5G2/ATP5G3 Recombinant Rabbit Monoclonal Antibody (K01_1N27)
GREENER_CHOICE

Produktkod. 30826983
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30826983 100 μL
30827449 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30826983 Leverantör Invitrogen™ Leverantörsnummer MA563263

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ARNPSLKQQL FSY.

The ATP5MC1 gene is one of three human paralogs that encode membrane subunit c of the mitochondrial ATP synthase. It is mapped to 17q21.32. This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene is one of three genes that encode subunit c of the proton channel. Each of the three genes have distinct mitochondrial import sequences but encode the identical mature protein. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen ATP5G1/ATP5G2/ATP5G3
Användningsområden Immunohistochemistry (Paraffin), Western Blot
Klassificering Recombinant Monoclonal
Klona K01_1N27
Koncentration 0.60 mg/mL
Konjugera Unconjugated
Formulering 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gen ATP5G1
Genaccessionsnr. P05496, P48201, Q06055, Q06646
Gene Alias 1810041M08Rik; 6030447M23; ATP synthase c subunit; ATP synthase F(0) complex subunit C1, mitochondrial; ATP synthase F(0) complex subunit C2, mitochondrial; ATP synthase F(0) complex subunit C3, mitochondrial; ATP synthase H+ transporting mitochondrial F0 complex subunit c (subunit 9) isoform 1; ATP synthase lipid-binding protein; ATP synthase lipid-binding protein, mitochondrial; ATP synthase membrane subunit c locus 1; ATP synthase membrane subunit c locus 2; ATP synthase membrane subunit c locus 3; ATP synthase proteolipid P1; ATP synthase proteolipid P2; ATP synthase proteolipid P3; ATP synthase proton-transporting mitochondrial F(0) complex subunit C1; ATP synthase proton-transporting mitochondrial F(0) complex subunit C2; ATP synthase proton-transporting mitochondrial F(0) complex subunit C3; ATP synthase subunit 9; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9) isoform 3; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 3; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c, isoform 3; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C3 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C3 (subunit 9); ATP synthase, mitochondrial, C subunit-3; ATP5A; ATP5G; ATP5G1; ATP5G2; Atp5g3; ATP5MC1; Atp5mc2; ATP5MC3; ATPase protein 9; ATPase subunit 9; ATPase subunit c; mitochondrial ATP synthase, subunit 9, isoform 1; mitochondrial ATP synthase, subunit C (subunit 9), isoform 2; mitochondrial ATP synthase, subunit C, isoform 1; P3; PSEC0033
Gensymboler ATP5G1, ATP5G2, ATP5G3
Värd art Rabbit
Immunogen A synthetic peptide of human ATP5G1/G2/G3 (aa 98-110).
Reningsmetod Protein A/G
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 171082, 516, 517, 518
Målarter Human, Rat
Innehåll och lagring Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.