Learn More
Abnova™ ATP5G1 (Human) Recombinant Protein
Human ATP5G1 full-length ORF ( AAH04963, 18 a.a. - 136 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH04963 |
---|---|
Gene ID (Entrez) | 516 |
Name | ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9) |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16162051
|
Abnova™
H00000516-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16172051
|
Abnova™
H00000516-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: TRGLIRPVSASFLSSPVNSSKQPSYSNFPLQVARREFQTSVVSRDIDTAAKFIGAGAATVGVAGSGAGIGTVFGSLIIGYARNPSLKQQLFSYAILGFALSEAMGLFCLMVAFLILFAM
Specifications
AAH04963 | |
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9) | |
125% SDS-PAGE Stained with Coomassie Blue | |
ATP5A, ATP5G | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
516 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP5G1 | |
GST |