missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15097-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
ATP6V1B2 is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A, three B, and two G subunits, as well as a C, D, E, F, and H subunit. The V1 domain contains the ATP catalytic site. ATP6V1B2 is one of two V1 domain B subunit isoforms and is the only B isoform highly expressed in osteoclasts.Specifications
ATP6V1B2 | |
Polyclonal | |
Unconjugated | |
ATP6V1B2 | |
ATP6B1B2, ATP6B2, ATP6V1B2, HO57, V ATPase subunit B 2, VATB, Vma2, VPP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117596, 11966, 526 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21281, P62814, P62815 | |
ATP6V1B2 | |
ATP6V1B2 Fusion Protein Ag7174 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |