missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPAF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11598-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 alpha subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. This gene is located within the Smith-Magenis syndrome region on chromosome 17. An alternatively spliced transcript variant has been described, but its biological validity has not been determined.Specifications
ATPAF2 | |
Polyclonal | |
Unconjugated | |
ATPAF2 | |
ATP12, ATP12 homolog, ATP12p, ATPAF2, LP3663 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246782, 303190, 91647 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N5M1, Q91YY4 | |
Atpaf2 | |
ATPAF2 Fusion Protein Ag2164 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |