missing translation for 'onlineSavingsMsg'
Learn More

ATPIF1 Recombinant Protein, Abnova

Brand:  Abnova H00093974-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16176853

  • 3343.96 SEK / 10µg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH09677
Solution
93974
ATPIF1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
ATPI/ATPIP/IP/MGC1167/MGC8898
ATPIF1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
37.8
8
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATPIF1
Human
Yes
Videos
SDS
Documents
Certificates