Läs mer
Atracurium Besylate (cis-Quaternary Alcohol), TRC
Produktkod. 30344069
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30344069 5 mg
30333484 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30344069 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7944205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Atracurium Besylate (cis-Quaternary Alcohol)
CAS 1100676-16-1
Molekylformel C29 H42 N O7 . C6 H5 O3 S
Kvantitet 5 mg
Synonym Isoquinolinium, 1-[(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-2-[3-[(5-hydroxypentyl)oxy]-3-oxopropyl]-6,7-dimethoxy-2-methyl-, (1R,2R)-, benzenesulfonate (1:1)
InChI formel InChI=1S/C29H42NO7.C6H6O3S/c1-30(14-12-29(32)37-16-8-6-7-15-31)13-11-22-19-27(35-4)28(36-5)20-23(22)24(30)17-21-9-10-25(33-2)26(18-21)34-3;7-10(8,9)6-4-2-1-3-5-6/h9-10,18-20,24,31H,6-8,11-17H2,1-5H3;1-5H,(H,7,8,9)/q+1;/p-1/t24-,30-;/m1./s1
LEDER COc1ccc(C[C@@H]2c3cc(OC)c(OC)cc3CC[N@+]2(C)CCC(=O)OCCCCCO)cc1OC.[O-]S(=O)(=O)c4ccccc4
IUPAC-namn benzenesulfonate;5-hydroxypentyl 3-[(1R,2R)-1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propanoate
Molekylvikt (g/mol) 673.813
Formel vikt 673.292