Läs mer
Atrazine Glutathione Adduct, TRC
Produktkod. 30329137
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30329137 1 mg
1 options
Produktkod. 30329137 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7946101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Atrazine Glutathione Adduct
CAS 24429-05-8
Molekylformel C18H30N8O6S
Kvantitet 1 mg
Synonym L-γ-Glutamyl-S-[4-(ethylamino)-6-[(1-methylethyl)amino]-1,3,5-triazin-2-yl]-L-cysteinylglycine, S-(4-Ethylamino-6-isopropylamino-2-s-triazino)glutathione, Glutathione S-Atrazine, ATR-SG
InChI formel InChI=1S/C18H30N8O6S/c1-4-20-16-24-17(22-9(2)3)26-18(25-16)33-8-11(14(30)21-7-13(28)29)23-12(27)6-5-10(19)15(31)32/h9-11H,4-8,19H2,1-3H3,(H,21,30)(H,23,27)(H,28,29)(H,31,32)(H2,20,22,24,25,26)/t10-,11-/m0/s1
LEDER CCNC1=NC(=NC(=N1)SC[C@@H](C(=O)NCC(=O)O)NC(=O)CC[C@@H](C(=O)O)N)NC(C)C
IUPAC-namn (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-[[4-(ethylamino)-6-(propan-2-ylamino)-1,3,5-triazin-2-yl]sulfanyl]-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
Molekylvikt (g/mol) 486.55