Läs mer
Atrazine-hydroxydesethyl, TRC
Produktkod. 30330189
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30330189 100 mg
1 options
Produktkod. 30330189 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA794605100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Amino-2-hydroxy-6-(isopropylamino)-1,3,5-triazine
CAS 19988-24-0
Molekylformel C6 H11 N5 O
Kvantitet 100 mg
Synonym s-Triazin-2-ol, 4-amino-6-(isopropylamino)- (8CI), 4-Amino-6-[(1-methylethyl)amino]-1,3,5-triazin-2(1H)-one, 2-Hydroxy-4-amino-6-(isopropylamino)-s-triazine, 2-Hydroxydesethylatrazine, 4-Amino-2-hydroxy-6-(isopropylamino)-1,3,5-triazine, Atrazine-hydroxydesethyl, GS 17794, Isopropylammeline, N-Isopropylammeline
InChI formel InChI=1S/C6H11N5O/c1-3(2)8-5-9-4(7)10-6(12)11-5/h3H,1-2H3,(H4,7,8,9,10,11,12)
LEDER CC(C)Nc1nc(N)nc(O)n1
IUPAC-namn 4-amino-6-(propan-2-ylamino)-1,3,5-triazin-2-ol
Molekylvikt (g/mol) 169.18
Formel vikt 169.0964