Läs mer
(±)-Atropine-d3 (N-methyl-d3), CDN
Specifikationer
Specifikationer
| Kemiskt namn eller material | Atropine D3 (N-methyl D3) |
| CAS | 60365-55-1 |
| Molekylformel | C17 D3 H20 N O3 |
| Kvantitet | 0.01 g |
| Synonym | Benzeneacetic acid, α-(hydroxymethyl)-, 8-(methyl-d3)-8-azabicyclo[3.2.1 ]oct-3-yl ester, endo- (9 CI), Benzeneacetic acid, α-(hydroxymethyl)-, 8-(methyl-d3)-8-azabicyclo[3.2.1 ]oct-3-yl ester, endo-(±)- (ZCI), α-(Hydroxymethyl)benzeneacetic acid endo-8-(methyl-d3)-8-azabicyclo[3.2.1 ]oct-3-yl ester, Atropine-(N-methyl-D3), Atropine D3 (N-methyl D3), (±)-Atropine-D3, (±)-Atropine-D3 |
| InChI formel | InChI=1 S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19 H,7-11H2,1H3/t13-,14+,15+,16-/m0/s1/i1D3 |
| LEDER | [2 H]C([2 H])([2 H])N1[C@@H]2 CC[C@H]1 C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3 |
| IUPAC-namn | [rac-(1 R,5 S)-8-(trideuteriomethyl)-8-azabicyclo[3.2.1 ]octan-3-yl] rac-(2 R)-3-hydroxy-2-phenylpropanoate |
| Molekylvikt (g/mol) | 292.3879 |
| Formel vikt | 292.1866 g/mol |
| Visa mer |