Läs mer
Atropine-N-oxide Hydrochloride, TRC
Produktkod. 30134019
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134019 500 mg
30136186 100 mg
30009688 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134019 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA793860500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4574-60-1
Molekylformel C17 H23 N O4 . Cl H
Molekylvikt (g/mol) 341.83
Synonym Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-oxido-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (1:1), 1αH,5αH-Tropan-3α-ol, (±)-tropate (ester), 8-oxide, hydrochloride (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-oxido-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (9CI), Benzeneacetic acid, α-(hydroxymethyl)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester endo-(±)-, N-oxide, hydrochloride, Atropine N-oxide hydrochloride, Atropine aminoxide hydrochloride, Atropine oxide hydrochloride, Genatropine hydrochloride, Tropinox, Xtro
IUPAC-namn [(1R,5S)-8-methyl-8-oxido-8-azoniabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate;hydrochloride
LEDER Cl.C[N+]1([O-])[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3
Specifikationer
Kemiskt namn eller material Atropine N-Oxide Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C17H23NO4.ClH/c1-18(21)13-7-8-14(18)10-15(9-13)22-17(20)16(11-19)12-5-3-2-4-6-12;/h2-6,13-16,19H,7-11H2,1H3;1H/t13-,14+,15+,16-,18?;/m0./s1
Formel vikt 341.1394
Procent renhet >95%