Learn More
Attune™ NxT Dichroic Filter, 600DLP
This is a replacement 600 DLP dichroic filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
Brand:  Invitrogen 100022771
Additional Details : Weight : 0.10000kg
Product Code. 15340657
  • 3025.00 SEK / Each
Estimated Shipment: 16-04-2024
to see stock.
Description
Description
  • Replacement dichroic filter
  • Used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer
Specifications
Specifications
Attune Acoustic Focusing Cytometer
Store at room temperature.
Dichroic Filter
Attune™