Learn More
Attune™ NxT Dichroic Filter, 600DLP
This is a replacement 600 DLP dichroic filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
Brand: Invitrogen 100022771
Additional Details : Weight : 0.10000kg
Description
- Replacement dichroic filter
- Used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer
Specifications
Attune Acoustic Focusing Cytometer | |
Store at room temperature. | |
Dichroic Filter | |
Attune™ |
For Research Use Only. Not for use in diagnostic procedures.