missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15693-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The autosomal domit cerebellar ataxias (ADCAs) are a clinically and genetically heterogeneous group of disorders characterized by ataxia, dysarthria, dysmetria, and intention tremor. All ADCAs involve some degree of cerebellar dysfunction and a varying degree of signs from other components of the nervous system. A commonly accepted clinical classification (Harding, 1993) divides ADCAs into 3 different groups based on the presence or absence of associated symptoms such as brainstem signs or retinopathy. The presence of pyramidal and extrapyramidal symptoms and ophthalmoplegia makes the diagnosis of ADCA I, the presence of retinopathy points to ADCA II, and the absence of associated signs to ADCA III. Genetic linkage and molecular analyses revealed that ADCAs are genetically heterogeneous even within the various subtypes.Specifications
ATXN10 | |
Polyclonal | |
Unconjugated | |
ATXN10 | |
ataxin 10, ATXN10, Brain protein E46 homolog, E46L, SCA10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170821, 25814, 54138 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P28658, Q9ER24, Q9UBB4 | |
Atxn10 | |
ATXN10 Fusion Protein Ag8259 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |