Läs mer
Augmentin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Augmentin |
| CAS | 74469-00-4 |
| Molekylformel | 4(C16H19N3O5S)•(C8H8KNO5) |
| Kvantitet | 1 g |
| Synonym | Mixture of Amoxicillin and Potassium Clavulanate (~4:1), Amocla, Amoclan, Amoclav, Amoran, Amox-clav, Amoxicillin-clavulanate Potassium Mixture, Amoxicillin-potassium Clavulanate Mixture, Amoxyclav, Ancla, Augmentan, Augmentin, Augmentin (antibiotic), Augmentin ES 600, Augmentine, Auspilic, BRL 25000, BRL 25000A, BRL 25000G, Clamentin, Clamobit, Clamonex, Clamoxyl, Clavamox, Clavinex, Clavoxilin Plus, Clavulin, Clavumox, Eumetinex, Forcid Solutab, Kmoxilin, Proclav S, Spectramox, Viaclav, Xiclav, co-Amoxyclav, Potassium (2R,5R,Z)-3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate (2S,5R,6R)-6-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Mixture |
| InChI formel | InChI=1S/C16H19N3O5S.C8H9NO5.K/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7;10-2-1-4-7(8(12)13)9-5(11)3-6(9)14-4;/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24);1,6-7,10H,2-3H2,(H,12,13);/q;;+1/p-1/b;4-1-;/t9-,10-,11+,14-;6-,7-;/m11./s1 |
| LEDER | O=C1N([C@H]/2C(O[K])=O)[C@](C1)([H])OC2=C/CO.O=C3N4[C@]([C@@H]3NC([C@H](N)C5=CC=C(O)C=C5)=O)([H])SC(C)(C)[C@@H]4C(O)=O |
| IUPAC-namn | potassium;(2S,5R,6R)-6-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;(2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| Molekylvikt (g/mol) | 4365.4023725 |
| Formel vikt | 602.1085 |