Beskrivning
- Original 3-panel design allows for greater facial movement
- Low-resistance filter technology helps you breathe more easily
- Sculpted upper panel conforms to the nose and contours of the face for an effective seal
- Embossed top panel helps reduce eyewear fogging
- Chin tab designed for ease of positioning and adjustment
- Easy to store away with flat-fold design and individual wrapping
- Colour-coded headbands for easy identification: yellow for FFP1 (NFP 4)
- Sculpted nose panel helps conform to the nose and contours of the face and helps to improve compatibility with 3M eyewear
- Maximum usage level: Up to 4 x TLV
- 3M™ Cool Flow™ Valve helps reduce heat build-up in hot and humid conditions
- Available in large or small pack sizes
Specifikationer
Specifikationer
| Tilldelad skyddsfaktor (APF) | FFP1 |
| Beskrivning | 9310+ |
| Produkttyp | Particulate Respirator |
| Med utandningsventil | No |