Läs mer
Auranofin, MedChemExpress
Produktkod. 16488668
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
Förpackningsstorlek:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16488668 10 mg 10mg
16404557 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16488668 Leverantör Medchem Express Leverantörsnummer HYB1123
MedChemExpress Auranofin (SKF-39162) is a thioredoxin reductase (TrxR) inhibitor with an IC50 of 0.2 μM. Auranofin exhibits antiviral activity against SARS-CoV21, with a CC50 of 4.2 μM for monkey kidney Vero E6 cells.
Auranofin (SKF-39162) is a thioredoxin reductase (TrxR) inhibitor with an IC50 of 0.2 μM. Auranofin exhibits antiviral activity against SARS-CoV21, with a CC50 of 4.2 μM for monkey kidney Vero E6 cells.
Specifikationer
Kemiskt namn eller material Auranofin
Fysisk form Powder
CAS 34031-32-8
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C20H36AuO9PS
För användning med (applikation) COVID-19-immunoregulation
Synonym SKF-39162
Löslighetsinformation DMSO : 50 mg/mL (73.48 mM; Need ultrasonic)
LEDER CC(O[C@@H]1[C@@H](OC(C)=O)[C@H]([SH-][Au+]P(CC)(CC)CC)O[C@H](COC(C)=O)[C@H]1OC(C)=O)=O
Molekylvikt (g/mol) 680.5
Formel vikt 680.5
Procent renhet 98.0%