Läs mer
Auranofin, TRC
Produktkod. 30332459
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30332459 50 mg
1 options
Produktkod. 30332459 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79475050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Auranofin
CAS 34031-32-8
Molekylformel C20 H34 Au O9 P S
Kvantitet 50 mg
Synonym Gold, [1-(thio-κS)-β-D-glucopyranose 2,3,4,6-tetraacetato](triethylphosphine)-, Gold, (1-thio-β-D-glucopyranosato)(triethylphosphine)-, 2,3,4,6-tetraacetate (8CI), Gold, (1-thio-β-D-glucopyranose 2,3,4,6-tetraacetato-S)(triethylphosphine)-, [1-(Thio-κS)-β-D-glucopyranose 2,3,4,6-tetraacetato](triethylphosphine)gold, β-D-Glucopyranose, 1-thio-, 2,3,4,6-tetraacetate, gold complex, Aktil, Auranofin, Crisinor, Crisofin, NSC 321521, Ridaura, Ridauran, SKF 39162, SKF 39162D, [(Tetra-O-acetyl-β-D-glucopyranosyl)thio](triethylphosphine)gold
InChI formel InChI=1S/C14H20O9S.C6H15P.Au/c1-6(15)19-5-10-11(20-7(2)16)12(21-8(3)17)13(14(24)23-10)22-9(4)18;1-4-7(5-2)6-3;/h10-14,24H,5H2,1-4H3;4-6H2,1-3H3;/q;;+1/p-1/t10-,11-,12+,13-,14+;;/m1../s1
LEDER CCP(=[Au]S[C@@H]1O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C)(CC)CC
IUPAC-namn [(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]sulfanyl-(triethyl-λ^{5}-phosphanylidene)gold
Molekylvikt (g/mol) 678.48
Formel vikt 678.1327