missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 4431-00-9 |
| Color | Maroon |
| Molecular Formula | C22H16O9 |
| Quantity | 10 g |
| Synonym | aurintricarboxylic acid, aluminon free acid, aluminon, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dienylidenemethylene di salicylic acid, benzoic acid, 5-3-carboxy-4-hydroxyphenyl 3-carboxy-4-oxo-2,5-cyclohexadien-1-ylidene methyl-2-hydroxy, 1,4-cyclohexadiene-1-carboxylic acid, 3-bis 3-carboxy-4-hydroxyphenyl methylene-6-oxo, 3,3'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 6-hydroxybenzoic acid, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 2-hydroxybenzoic acid |
| InChI Key | YETAHXYTPZCNBG-UHFFFAOYNA-N |
| SMILES | OC(=O)C1=CC(C=CC1=O)C(C1=CC=C(O)C(=C1)C(O)=O)C1=CC=C(O)C(=C1)C(O)=O |
| IUPAC Name | 5-[(3-carboxy-4-hydroxyphenyl)(3-carboxy-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoic acid |
| Molecular Weight (g/mol) | 424.36 |
| PubChem CID | 2259 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?