Läs mer
Auristatin F TFA Salt, TRC
Produktkod. 30347264
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30347264 1 mg
30341191 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30347264 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7941001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Auristatin F TFA Salt
CAS 163768-50-1
Molekylformel C40H67N5O8 • x(C2HF3O2)
Kvantitet 1 mg
Synonym N,N-Dimethyl-L-valyl-L-valyl-(3R,4S,5S)-3-methoxy-5-methyl-4-(methylamino)heptanoyl-(αR,βR,2S)-β-methoxy-α-methyl-2-pyrrolidinepropanoyl-L-phenylalanine TFA Salt
InChI formel InChI=1S/C40H67N5O8/c1-13-26(6)35(44(10)39(49)33(24(2)3)42-38(48)34(25(4)5)43(8)9)31(52-11)23-32(46)45-21-17-20-30(45)36(53-12)27(7)37(47)41-29(40(50)51)22-28-18-15-14-16-19-28/h14-16,18-19,24-27,29-31,33-36H,13,17,20-23H2,1-12H3,(H,41,47)(H,42,48)(H,50,51)/t26-,27+,29-,30-,31+,33-,34-,35-,36+/m0/s1
LEDER O=C(N[C@H](C(C)C)C(N(C)[C@H](C(CC)C)[C@H](OC)CC(N1CCC[C@@]1([H])[C@@H](OC)[C@@H](C)C(N[C@@H](C(O)=O)CC2=CC=CC=C2)=O)=O)=O)[C@@H](N(C)C)C(C)C
IUPAC-namn (2S)-2-[[(2R,3R)-3-[(2S)-1-[(3R,4S,5S)-4-[[(2S)-2-[[(2S)-2-(dimethylamino)-3-methylbutanoyl]amino]-3-methylbutanoyl]-methylamino]-3-methoxy-5-methylheptanoyl]pyrrolidin-2-yl]-3-methoxy-2-methylpropanoyl]amino]-3-phenylpropanoic acid
Molekylvikt (g/mol) 745.9911402