Läs mer
Tocris Bioscience™ AUTAC4
Beskrivning
AUTAC4 is an autophagy-targeting chimera targeting mitochondria comprising p-fluorobenzyl guanine joined by a PEG linker to a 2-phenylindole-3-glyoxyaminde ligand for the translocator protein on the outer mitochondrial membrane. It delivers a guanine tag to the mitochondrial membrane and promotes mitophagy of damaged mitochondria. In Down syndrome cells, AUTAC4 degrades impaired mitochondria and promotes the generation of functionally normal mitochondria.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[(16 R)-16-[[[2-Amino-9-[(4-fluorophenyl)methyl]-6,9-dihydro-6-oxo-1 H-purin-8-yl]thio]methyl]-15,18-dioxo-4,7,10-trioxa-14,17-diazanonadec-1-yl]-α-oxo-2-phenyl-1 H-indole-3-acetamide |
| CAS | 2267315-04-6 |
| Kvantitet | 5 mg |
| Mål | Active Degraders |
| Molekylformel | C43H48FN9O8S |
| Renhet | 0.98 |