Läs mer
MedChemExpress Autocamtide 2
Produktkod. 16482689
Encompass_Preferred
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
16482689 1 mg
1 options
Produktkod. 16482689 Leverantör MedChemExpress Leverantörsnummer HYP0225
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
MedChemExpress Autocamtide 2 is a highly selective peptide substrate of calcium/calmodulin-dependent protein kinase II (CaMKII). It can be used in the CaMKII activity assay.
Specifikationer
Renhet 98.21%
CAS 129198-88-5
Innehåll och lagring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
För användning med (applikation) Neuroscience-Neuromodulation
Molekylvikt (g/mol) 1527.77 g/mol
Produkttyp Peptides
Kvantitet 1 mg
Frakt skick Room temperature
Löslighet H2O : 20 mg/mL (13.09 mM∣Need ultrasonic)
Synonym Autocamtide II