Learn More
Abeba™ Autoclavable Clogs 9600
Antistatic and autoclavable at 121°C. High slip resistance.
Marca: Abeba™ 9600/41
Detalles adicionales : Weight : 0.50000kg
Descripción
- CE EN ISO 20347:2012 OB A, E SRC
- Autoclavable, sterilisable special material
- White
- Slip resistance outsole sole
- Antistatic
- Heel strap foldable, adjustable
- DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Especificaciones
CE EN ISO 20347:2012 OB A, E SRC | |
41 | |
9600 |
White | |
Slip-resistant |