Learn More
Abeba™ Autoclavable Clogs 9600
Antistatic and autoclavable at 121°C. High slip resistance.
Brand:  Abeba™ 9600/43
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 15539090
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • White
  • Slip resistance outsole sole
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRC
43
9600
White
Slip-resistant