Learn More
Poulten Graf™ Autoclavable Glass Syringe
Able to be autoclaved at 134°C. Poulten Graf™ Autoclavable Glass Syringes come with an interchangeable piston.
Brand: Poulten Graf™ 7 10221
Additional Details : Weight : 0.01170kg
Description
- Complies with CE EN 46001 standards
- Constructed from Durobax™ glass
- With permanent amber graduation
- Acid and alkali resistant
Specifications
Autoclavable | |
Syringe | |
1 mL |
Luer Tip | |
1/Ea. |