Learn More
ATAGO™ Automatic Digital Refractometer RX-007α
Product Code. 10141123
Change view
Click to view available options
Dimensions (L x W x H):
370 x 260 x 140 mm
Unit Size:
Each
Product Code. Dimensions (L x W x H) Unit Size
10141123 370 x 260 x 140 mm Each
1 options
Product Code. 10141123 Supplier ATAGO™ Supplier No. 3921
  • ONLINE EXCLUSIVE PRICE
    79095.00 SEK
    63061.47 SEK / Paket med 1
    Save 16033.53 SEK
    20% Av
Beräknad leverans: 28-12-2026
The Automatic Digital Refractometer RX-007α is suitable for measuring water soluble samples with very low concentrations, such as tea and low sugar beverages, at a very high accuracy.
  • The new RX-007α (alpha) is suitable for measuring water soluble samples with very low concentrations (5.000% or less) such as tea (green tea, black tea, oolong tea) and low sugar beverages at a very high accuracy of ±0.005%.
  • This product comes standard with a two (2) year limited warranty against manufacturer's defects from the date of the original purchase. This warranty does not cover or apply to the prism. The warranty period can be extended to three (3) years if the product is registered with ATAGO.
  • FDA 21 CFR Part 11 Software Included in Standard Delivery.
Specifications
Type Benchtop Refractometer
Refractive Index 1.330150 to 1.341500
Accuracy ±0.005% (Brix), 0.000010 (Refractive Index)
Resolution 0.001% (Brix), 0.000001 (Refractive Index)
Weight (Metric) 6.7 kg
Dimensions (L x W x H) 370 x 260 x 140 mm
Temperature Accuracy 0.001°C
Temperature Range (Metric) 15.00°C to 30.00°C