Learn More
Trajan Scientific and Medical™ Autosampler Syringes for Agilent GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
Brand: Trajan Scientific and Medical™ 002826
Additional Details : Weight : 0.10000kg
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Agilent instruments
Specifications
Agilent Syringe | |
0.11 mm | |
Fixed Needle with GT Plunger | |
Cone |
23 and 26 G | |
0.63, 0.47 mm | |
10 μL |